missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma R2 Antibody (007), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90670AF594
This item is not returnable.
View return policy
Description
IFN-gamma R2 Monoclonal specifically detects IFN-gamma R2 in Mouse samples. It is validated for ELISA.
Specifications
| IFN-gamma R2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFN-gamma-R2, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFN-gamma R2 (NP_032364.1; Met1-Val243). | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse IFNGR1 Mouse IFNGR1 Human IL10RB | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 7 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3460 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction