missing translation for 'onlineSavingsMsg'
Learn More

IFN-gamma R1/CD119 Antibody (062), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18336250 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336250 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336250 Supplier Novus Biologicals Supplier No. NBP290685AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-gamma R1/CD119
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 62
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFN-gamma-R1, IFNGR, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFN-gamma R1/CD119 (P15261; Met1-Asp253).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Innate Immunity, Myeloid derived Suppressor Cell, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3459
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.