missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IFN-gamma Antibody Pair

Product Code. 30042582
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 30042582

Brand: Novus Biologicals™ NBP33075715Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25-2000 pg/mL (example only; lot dependent)
Assay Sensitivity 31.25 pg/mL (example only; lot dependent)
Conjugate Unconjugated
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Mouse IFN-gamma.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3458
Gene Symbol IFNG
Quantity 15 Plates
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Target Mouse
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.