missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-gamma Antibody (AMG 811), DyLight 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-28004UV
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal antibody specifically detects IFN-gamma in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IFN-gamma | |
| Monoclonal | |
| DyLight 350 | |
| IFG, IFI, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| IFNg | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Stem Cell Markers | |
| 3458 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| AMG 811 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction