missing translation for 'onlineSavingsMsg'
Learn More

IFN-alpha WA/IFNA16 Antibody, Novus Biologicals™

Product Code. 18359688 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359688 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359688 Supplier Novus Biologicals Supplier No. H00003449D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFN-alpha WA/IFNA16 Polyclonal antibody specifically detects IFN-alpha WA/IFNA16 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-alpha WA/IFNA16
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_002164.1
Gene Alias IFN-alpha-16, IFN-alpha-N-protein, IFN-alphaO, interferon alpha-16, Interferon alpha-WA, interferon, alpha 16
Host Species Rabbit
Immunogen IFNA16 (NP_002164.1, 1 a.a. - 189 a.a.) full-length human protein. MALSFSLLMAVLVLSYKSICSLGCDLPQTHSLGNRRALILLAQMGRISHFSCLKDRYDFGFPQEVFDGNQFQKAQAISAFHEMIQQTFNLFSTKDSSAAWDETLLDKFYIELFQQLNDLEACVTQEVGVEEIALMNEDSILAVRKYFQRITLYLMGKKYSPCAWEVVRAEIMRSFSFSTNLQKGLRRKD
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3449
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.