missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN alpha Recombinant Superclonal™ Antibody (9HCLC)

Product Code. 15397294
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15397294 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15397294 Supplier Invitrogen™ Supplier No. 710299

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with non-human primate based on sequence homology. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN alpha
Applications Western Blot
Classification Recombinant Superclonal
Clone 9HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene IFNA1
Gene Accession No. P01562, P01563
Gene Alias alpha-2a interferon; Gm12597; ifa; Ifa1; Ifa2; IFL; IFN; IFN alpha; IFN alpha 1/13; IFN alpha 1b; IFN α; IFN1@; IFNA; IFNA@; IFNA1; IFNA13; Ifna15; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFNA3; IFNA6; Ifnaa; IFNAalphaIFN-alpha-1/13; IFNalpha; IFN-alpha; IFN-alpha 1b; IFN-alpha1; IFN-alpha-1; IFN-alpha-1/13; IFN-alpha-2; IFN-alpha-6; IFN-alphaA; IFN-alphaD; IFN-gamma; IFNα; INFA2; interferon; interferon alpha; interferon alpha 1; Interferon alpha 13 (IFNA13); interferon alpha 15; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family gene 1; interferon alpha family, gene 2; Interferon alpha leukocyte (IFL); interferon alpha protein; interferon alpha-1; Interferon alpha1 (IFNA1); interferon alpha-1 precursor; interferon alpha-1/13; Interferon alpha-1/13 (IFN-alpha-1/13); Interferon alpha-2; Interferon alpha-A; Interferon alpha-D; Interferon alpha-D (IFN alphaD); interferon type A1/A2; interferon type A3; interferon, alpha 1; interferon, alpha 2; interferon-alpha; interferon-alpha 1; interferon-alpha1; interferon-alpha-6; leIF A; LeIF D; MGC138207; MGC138505; MGC138507; MuIFN-alpha-A; novel interferon alpha (Ifna) family member; OTTMUSG00000007655
Gene Symbols IFNA1, IFNA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 24-189 of human Interferon-alpha.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3439, 3440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.