missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IFN-alpha H2/IFNA14 Polyclonal specifically detects IFN-alpha H2/IFNA14 in Mouse samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IFN-alpha H2/IFNA14 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | IFN-alpha-14, IFN-alphaH, interferon alpha-14, Interferon alpha-H, Interferon lambda-2-H, interferon, alpha 14, LeIF H, LEIF2H, MGC125756, MGC125757 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Mouse IFN-alpha H2/IFNA14 (NP_996858.1; Cys24-Lys189). |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?