missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha H2/IFNA14 Antibody (065), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90671CL1
This item is not returnable.
View return policy
Description
IFN-alpha H2/IFNA14 Monoclonal antibody specifically detects IFN-alpha H2/IFNA14 in Mouse samples. It is validated for ELISA
Specifications
| IFN-alpha H2/IFNA14 | |
| Monoclonal | |
| CoraFluor 1 | |
| IFN-alpha-14, IFN-alphaH, interferon alpha-14, Interferon alpha-H, Interferon lambda-2-H, interferon, alpha 14, LeIF H, LEIF2H, MGC125756, MGC125757 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IFN-alpha H2/IFNA14 (Accession#: NP_996858.1; Cys24-Lys189). | |
| 0.1 mL | |
| Cytokine Research | |
| 3448 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 065 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction