missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha C/IFN-alpha 10 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | IFN-alpha C/IFN-alpha 10 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18351306
|
Novus Biologicals
NBP3-15494-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18372745
|
Novus Biologicals
NBP3-15494-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-alpha C/IFN-alpha 10 Polyclonal antibody specifically detects IFN-alpha C/IFN-alpha 10 in Rat samples. It is validated for Western BlotSpecifications
| IFN-alpha C/IFN-alpha 10 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| IFN-alpha-10, IFN-alphaC, interferon alpha-10, Interferon alpha-6L, Interferon alpha-C, interferon, alpha 10, LeIF C, MGC119878, MGC119879 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 24-100 of human IFN-alpha C/IFN-alpha 10 (NP_002162.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Immunology, Innate Immunity | |
| PBS, 50% glycerol, pH7.3 | |
| 3446 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title