missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IFN-alpha/beta R2 Monoclonal specifically detects IFN-alpha/beta R2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IFN-alpha/beta R2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 117 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFNARB, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 |
| Host Species | Rabbit |
| Immunogen | Recombinant Human IFN-alpha/beta R2 protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?