missing translation for 'onlineSavingsMsg'
Learn More

IFN-alpha/beta R2 Antibody (122), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18420738 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420738 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420738 Supplier Novus Biologicals Supplier No. NBP289447MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IFN-alpha/beta R2 Monoclonal antibody specifically detects IFN-alpha/beta R2 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-alpha/beta R2
Applications Flow Cytometry
Classification Monoclonal
Clone 122
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFNARB, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human IFN-alpha/beta R2 (NP_997468.1; Met1-Lys243).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3455
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.