missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IFN-alpha/beta R1 Antibody Pair [HRP]

Product Code. 18223808 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18223808

missing translation for 'mfr': Novus Biologicals™ NBP2794125PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IFN-alpha/beta R1.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78 pg/mL
Assay Sensitivity 78.13-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IFN-alpha/beta R1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1
Gene ID (Entrez) 3454
Gene Symbol IFNAR1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 95 ng of recombinant IFN-alpha/beta R1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-IFN-alpha/beta R1 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL mouse anti-IFN-alpha/beta R1 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-protector, pH 7.4).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Neuroscience, Phospho Specific
    Storage Requirements Storage is content dependent.
    Target IFN-alpha/beta R1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.