Learn More
IFN-alpha/beta R1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32196JF549
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Polyclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot
Specifications
| IFN-alpha/beta R1 | |
| Polyclonal | |
| Rabbit | |
| Immunogen affinity purified |
| Western Blot | |
| Janelia Fluor 549 | |
| The immunogen this antibody was made to, maps to a region between residue 507 to 557 of human Interferon (alpha, beta and omega) Receptor 1 using the numbering given in entry NP_000620.2 (GeneID 3454). | |
| 0.1 mL |