missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha/beta R1 Antibody (2951C), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB2452A
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| IFN-alpha/beta R1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 2951C | |
| Western Blot, Flow Cytometry | |
| alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
| Mouse myeloma cell line, NS0-derived human IFN-alpha/beta R1, Gly26-Lys436, Accession # P17181 | |
| 0.1 mL | |
| Neuroscience, Phospho Specific | |
| 3454 | |
| Store at 4C in the dark. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering