missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFN-alpha/beta R1 Antibody (1B12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26209-100ul
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Monoclonal antibody specifically detects IFN-alpha/beta R1 in Human samples. It is validated for ELISA
Specifications
| IFN-alpha/beta R1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1B12 | |
| ELISA | |
| alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFNBR, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
| A synthesized peptide derived from Human IFN-alpha/beta R1 [UniProt P17181] | |
| 100 μL | |
| Neuroscience, Phospho Specific | |
| 3454 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction