missing translation for 'onlineSavingsMsg'
Learn More

IFN-alpha 2 Antibody (ST29), PerCP, Novus Biologicals™

Product Code. 18407598 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407598 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407598 Supplier Novus Biologicals Supplier No. NB1002479PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Human samples. It is validated for ELISA, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-alpha 2
Applications ELISA, Functional Assay
Classification Monoclonal
Clone ST29
Conjugate PerCP
Dilution ELISA, Functional
Formulation PBS
Gene Alias alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765
Host Species Mouse
Immunogen Human interferon (HuIFN) produced by Namalwa cells (HuIFN-aN)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3440
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.