missing translation for 'onlineSavingsMsg'
Learn More

IFN-alpha 2 Antibody (N39), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18409234 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409234 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409234 Supplier Novus Biologicals Supplier No. NBP254398AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-alpha 2 Monoclonal antibody specifically detects IFN-alpha 2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-alpha 2
Applications Flow Cytometry, ELISA, Immunofluorescence, CyTOF
Classification Monoclonal
Clone N39
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias alpha-2a interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765
Host Species Mouse
Immunogen Purified recombinant human IFN-alpha 2 (Uniprot: P01563)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline BiologicAll speciesy Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 3440
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.