missing translation for 'onlineSavingsMsg'
Learn More

IFLTD1 Antibody - BSA Free, Novus Biologicals™

Product Code. 18331381 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331381 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18331381 Supplier Novus Biologicals Supplier No. NBP282095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFLTD1 Polyclonal specifically detects IFLTD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFLTD1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ36004, intermediate filament tail domain containing 1, intermediate filament tail domain-containing protein 1, Pas1c1
Gene Symbols IFLTD1
Host Species Rabbit
Immunogen IFLTD1 antibody was raised against an 19 amino acid synthetic peptide near the center of human IFLTD1. The immunogen is located within amino acids 200 - 250 of IFTLD1. Amino Acid Squence: EAKHQPPSDFLWKEQDKFR
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160492
Test Specificity At least four isoforms of IFLTD1 are known to exist; this antibody will detect all but isoform 4
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.