missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFITM3 Recombinant Rabbit Monoclonal Antibody (K01_1R35)
GREENER_CHOICE

Product Code. 30827126
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30827126 100 μL
30827297 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30827126 Supplier Invitrogen™ Supplier No. MA563747

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

IFITM3 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

IFITM3 (interferon induced transmembrane protein 3), also known as 1-8U or IP15, is a multi-pass membrane protein that belongs to the IFITM (interferon inducible transmembrane) family of proteins. IFITM proteins are induced by type I and type II interferons and contain multiple interferon (IFN)-stimulated response elements (ISREs) in their promoter regions. IFITM proteins play important roles in many cellular processes and their expression requires the presence of the chromatin remodeling SWI/SNF-like BAF complexes. Cellular processes involving IFITM proteins include cellular anti-proliferative activities and homotypic cell adhesion functions of interferons. In addition, IFITM genes are often upregulated in various cancer cells, suggesting a possible role in carcinogenesis. Localizing to the membrane, IFITM3 is a 133 amino acid protein that is induced by IFN-alpha and IFN-gamma. IFITM3 expression can be regulated by TEF-1, Brg-1 and Sp1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFITM3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1R35
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene IFITM3
Gene Accession No. Q01628
Gene Alias 1110004C05Rik; 1-8U; Cd225; Cdw217; Dispanin subfamily A member 2b; DSPA2b; Fgls; Fragilis; Fragilis 3; Fragilis protein; Ifitm3; ifitm-like protein 1; interferon induced transmembrane protein 3; interferon-induced transmembrane protein 3; interferon-inducible protein 15; Interferon-inducible protein 1-8U; interferon-inducible protein variant 10; IP15; mil-1; Mouse ifitm-like protein 1; rat8
Gene Symbols IFITM3
Host Species Rabbit
Immunogen A synthetic peptide of human Fragilis.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10410
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.