Learn More
Invitrogen™ IFITM3 Recombinant Rabbit Monoclonal Antibody (JU73-02)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532798
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
IFITM3 (interferon induced transmembrane protein 3), also known as 1-8U or IP15, is a multi-pass membrane protein that belongs to the IFITM (interferon inducible transmembrane) family of proteins. IFITM proteins are induced by type I and type II interferons and contain multiple interferon (IFN)-stimulated response elements (ISREs) in their promoter regions. IFITM proteins play important roles in many cellular processes and their expression requires the presence of the chromatin remodeling SWI/SNF-like BAF complexes. Cellular processes involving IFITM proteins include cellular anti-proliferative activities and homotypic cell adhesion functions of interferons. In addition, IFITM genes are often upregulated in various cancer cells, suggesting a possible role in carcinogenesis. Localizing to the membrane, IFITM3 is a 133 amino acid protein that is induced by IFN-alpha and IFN-gamma. IFITM3 expression can be regulated by TEF-1, Brg-1 and Sp1.
Specifications
| IFITM3 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| Q01628 | |
| IFITM3 | |
| Synthetic peptide within Human Fragilis aa 1-50. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| JU73-02 | |
| Unconjugated | |
| IFITM3 | |
| 1110004C05Rik; 1-8U; Cd225; Cdw217; Dispanin subfamily A member 2b; DSPA2b; Fgls; Fragilis; Fragilis 3; Fragilis protein; Ifitm3; ifitm-like protein 1; interferon induced transmembrane protein 3; interferon-induced transmembrane protein 3; interferon-inducible protein 15; Interferon-inducible protein 1-8U; interferon-inducible protein variant 10; IP15; mil-1; Mouse ifitm-like protein 1; rat8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10410 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.