missing translation for 'onlineSavingsMsg'
Learn More

IFITM2 Mouse anti-Human, Clone: 3D5F7, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66137-1-IG-20UL

 View more versions of this product

Product Code. 16817745

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific to IFTIM2.

IFN-induced antiviral protein which inhibits the entry of viruses to the host cell cytoplasm, permitting endocytosis, but preventing subsequent viral fusion and release of viral contents into the cytosol. Active against multiple viruses, including influenza A virus, SARS coronavirus (SARS-CoV), Marburg virus (MARV), Ebola virus (EBOV), Dengue virus (DNV), West Nile virus (WNV), human immunodeficiency virus type 1 (HIV-1) and vesicular stomatitis virus (VSV). Can inhibit: influenza virus hemagglutinin protein-mediated viral entry, MARV and EBOV GP1,2-mediated viral entry, SARS-CoV S protein-mediated viral entry and VSV G protein-mediated viral entry. Induces cell cycle arrest and mediates apoptosis by caspase activation and in p53-independent manner.
Specifications

Specifications

IFITM2
Monoclonal
0.88 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q01629
IFITM2
Protein G
RUO
10581
-20°C
Liquid
Western Blot, Flow Cytometry
3D5F7
Unconjugated
IFITM2
DSPA2c, IFITM2
Mouse
20 μL
Primary
Human
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.