Learn More
IFITM2/3 Mouse anti-Human, CoraLite 594, Clone: 2E8D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-66081-100UL
Description
The IFITM2/3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human IFITM2/3. This antibody recognizes human antigen. The IFITM2/3 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™594.
Specifications
| IFITM2/3 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 10410 | |
| IgG1 |
| Flow Cytometry (Intracellular Staining) | |
| 2E8D12 | |
| Mouse | |
| Primary | |
| Human |