missing translation for 'onlineSavingsMsg'
Learn More

IFIT5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£362.00

Specifications

Antigen IFIT5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18219264
View Documents
Novus Biologicals
NBP1-58887
100 μL
£362.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IFIT5 Polyclonal specifically detects IFIT5 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IFIT5
Polyclonal
Rabbit
Q13325
24138
Synthetic peptides corresponding to IFIT5(interferon-induced protein with tetratricopeptide repeats 5) The peptide sequence was selected from the middle region of IFIT5. Peptide sequence ITVYRLDDSDREGSVKSFSLGPLRKAVTLNPDNSYIKVFLALKLQDVHAE.
Primary
Western Blot
Unconjugated
RUO
FLJ53857, IFIT-5, interferon-induced protein with tetratricopeptide repeats 5, Retinoic acid- and interferon-inducible 58 kDa protein, retinoic acid- and interferon-inducible protein (58kD), RI58FLJ92678
IFIT5
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.