missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIT3 Antibody (OTI1G1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71006PE
This item is not returnable.
View return policy
Description
IFIT3 Monoclonal specifically detects IFIT3 in Human samples. It is validated for Flow Cytometry.
Specifications
| IFIT3 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| IFIT3 | |
| Full length recombinant protein of human IFIT3 (NP_001540) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI1G1 | |
| Flow Cytometry | |
| CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49 | |
| Mouse | |
| Affinity Purified | |
| Immunology | |
| 3437.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction