missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IFIT3 Antibody (2C10), Novus Biologicals™

Product Code. 18346498 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346498 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346498 Supplier Novus Biologicals™ Supplier No. H00003437M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFIT3 Monoclonal antibody specifically detects IFIT3 in Human samples. It is validated for ELISA,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT3
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2C10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001540
Gene Alias CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49
Host Species Mouse
Immunogen IFIT3 (NP_001540.2, 392 a.a. ∽ 490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. STDKEEIKDQPQNVSENLLPQNAPNYWYLQGLIHKQNGDLLQAAKCYEKELGRLLRDAPSGIGSIFLSASELEDGSEEMGQGAVSSSPRELLSNSEQLN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3437
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.