missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFIT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5148283
This item is not returnable.
View return policy
Description
IFIT1 Polyclonal antibody specifically detects IFIT1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)
Specifications
| IFIT1 | |
| Polyclonal | |
| Unconjugated | |
| IFIT1 | |
| C56; G10P1; Garg16; GARG-16; glucocorticoid-attenuated response gene 16 product; glucocorticoid-attenuated response gene 16 protein; Ifi56; IFI-56; IFI-56K; IFIT1; IFIT-1; IFNAI1; interferon induced protein with tetratricopeptide repeats 1; interferon, alpha-inducible protein (MW 56kD); interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-induced protein with tetratricopeptide repeats 1-like protein; interferon-inducible mRNA 561; Isg56; P56; RNM561 | |
| E. coli-derived Human IFIT1 fragment | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 100 μg/mL | |
| PBS with no preservative | |
| P09914 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3434 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction