missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFIT1 Monoclonal Antibody (OTI3G8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525050
This item is not returnable.
View return policy
Description
IFIT1 Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot
Interferon-induced antiviral RNA-binding protein that specifically binds single-stranded RNA bearing a 5'-triphosphate group (PPP-RNA), thereby acting as a sensor of viral single-stranded RNAs and inhibiting expression of viral messenger RNAs. Single-stranded PPP-RNAs, which lack 2'-O-methylation of the 5' cap and bear a 5'-triphosphate group instead, are specific from viruses, providing a molecular signature to distinguish between self and non-self mRNAs by the host during viral infection. Directly binds PPP-RNA in a non-sequence-specific manner. Viruses evolved several ways to evade this restriction system such as encoding their own 2'-O-methylase for their mRNAs or by stealing host cap containing the 2'-O-methylation (cap snatching mechanism). Exhibits antiviral activity against several viruses including human papilloma and hepatitis C viruses.
Specifications
| IFIT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09914 | |
| IFIT1 | |
| Full length human recombinant protein of IFIT1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI3G8 | |
| Unconjugated | |
| IFIT1 | |
| C56; G10P1; Garg16; GARG-16; glucocorticoid-attenuated response gene 16 product; glucocorticoid-attenuated response gene 16 protein; Ifi56; IFI-56; IFI-56K; IFIT1; IFIT-1; IFNAI1; interferon induced protein with tetratricopeptide repeats 1; interferon, alpha-inducible protein (MW 56kD); interferon-induced 56 kDa protein; Interferon-induced protein with tetratricopeptide repeats 1; interferon-induced protein with tetratricopeptide repeats 1-like protein; interferon-inducible mRNA 561; Isg56; P56; RNM561 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3434 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction