missing translation for 'onlineSavingsMsg'
Learn More

IFIT1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18409855 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409855 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409855 Supplier Novus Biologicals Supplier No. NBP281823AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFIT1 Polyclonal antibody specifically detects IFIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFIT1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias G10P1C56, GARG-16, IFI56IFI-56, IFI-56K, IFIT-1, IFNAI1ISG56, Interferon, alpha-inducible protein (MW 56kD), Interferon-induced 56 kDa protein, interferon-induced protein 56, interferon-induced protein with tetratricopeptide repeats 1, p56, RNM561
Host Species Rabbit
Immunogen IFIT1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human IFIT1. The immunogen is located within amino acids 290 - 340 of IFIT1. Amino Acid Squence: KEATKGQPRGQNREKLDK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3434
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.