missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550175
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total IFI6 protein.
G1P3, also known as Interferon alpha-inducible protein 6 (IFI6) and Interferon-induced protein 6-16, is a 130 amino acid member of the IFI6 family of proteins. Localized to the mitochondria, G1P3 is a multi-pass membrane protein that is induced by IFN-α2β. G1P3 has been shown to play a major role in the apoptosis pathway. Specifically, G1P3 acts as a cell survival protein by inhibiting caspase-3 activity, which antagonizes apoptosis. G1P3 has been implicated in tumorigenesis and is the subject of therapeutic studies, as reducing G1P3-mediated antiapoptotic signals could suggest improved therapies for myeloma or other malignancies.Specifications
IFI6 | |
Polyclonal | |
Unconjugated | |
IFI6 | |
6-16; FAM14C; G1P3; IFI6; IFI616; IFI-6-16; interferon alpha inducible protein 6; interferon alpha-inducible protein 6; interferon, alpha-inducible protein 6; interferon, alpha-inducible protein clone IFI-6-16; interferon-induced protein 6-16 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2537 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P09912 | |
IFI6 | |
Synthesized peptide derived from C-terminal of human IFI6. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |