missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI44L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38892
This item is not returnable.
View return policy
Description
IFI44L Polyclonal specifically detects IFI44L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| IFI44L | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Q53G44 | |
| IFI44L | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IIDEQLVCRLSKTDIFIICRDNKIYLDKMITRNLKLRFYGHRQYLECEVFRVEGIKDNLDDIKRIIKAREHRNRLL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| C1orf29, chromosome 1 open reading frame 29, GS3686, interferon-induced protein 44-like | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10964 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction