missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI44 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £417.00
Specifications
| Antigen | IFI44 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232692
|
Novus Biologicals
NBP3-38516-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230885
|
Novus Biologicals
NBP3-38516-20ul |
20 μL |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFI44 Polyclonal antibody specifically detects IFI44 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
| IFI44 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| interferon-induced protein 44, interferon-induced, hepatitis C-associated microtubular aggregate protein(44kD), MTAP44p44Microtubule-associated protein 44, P44 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of mouse IFI44 (NP_598632.2).,, Sequence:, FHKRKTNDFSILLDEKAVIVSSAICKMLQLTARNNVIPIQECEAFRCEELLDERKTRGIAVLHSNLLQALRDYKPYGDLVQQTRVLLLGPIGAGKSSFVNS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10561 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title