missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFI30 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5109845

Product Code. 17177773

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MTLSPLLLFL PPLLLLLDVP TAAVQASPLQ ALDFFGNGPP VNYKTGNLYL RGPLKKSNAP LVNVTLYYEA LCGGCRAFLI RELFPTWLLV MEILNVTLVP YGNAQEQNVS GRWEFKCQHG EEECKFNKVE ACVLDELDME LAFLTIVCME EFEDMERSLP LCLQLYAPGL SPDTIMECAM GDRGMQLMHA NAQRTDALQP PHEYVPWVTV NGKPLEDQTQ LLTLVCQLYQ GKKPDVCPSS TSSLRSVCFK.

The protein encoded by this gene is a lysosomal thiol reductase that at low pH can reduce protein disulfide bonds. The enzyme is expressed constitutively in antigen-presenting cells and induced by gamma-interferon in other cell types. This enzyme has an important role in MHC class II-restricted antigen processing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFI30
Polyclonal
Unconjugated
IFI30
Gamma-interferon-inducible lysosomal thiol reductase; gamma-interferon-inducible protein IP-30; GILT; IFI30; IFI-30; IFI30, lysosomal thiol reductase; interferon gamma inducible protein 30; interferon gamma-inducible protein 30 preproprotein; interferon, gamma-inducible protein 30; IP30; IP-30; Legumaturain
Rabbit
Affinity Chromatography
RUO
10437
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.58 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
P13284
IFI30
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human IFI30 (NP_006323.2).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.