missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI27L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69301
This item is not returnable.
View return policy
Description
IFI27L2 Polyclonal specifically detects IFI27L2 in Human samples. It is validated for Western Blot.
Specifications
| IFI27L2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FAM14A, family with sequence similarity 14, member A, interferon alpha-inducible protein 27-like protein 2, interferon, alpha-inducible protein 27-like 2, Interferon-stimulated gene 12b protein, ISG12(b), ISG12(b) protein, ISG12B, pIFI27-like protein, Protein TLH29, TLH29MGC44913 | |
| Rabbit | |
| 12 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9H2X8 | |
| IFI27L2 | |
| Synthetic peptides corresponding to FAM14A(family with sequence similarity 14, member A) The peptide sequence was selected from the middle region of FAM14A. Peptide sequence LSTSSNILLASVGSVLGACLGNSPSSSLPAEPEAKEDEARENVPQGEPPK. | |
| Affinity purified | |
| RUO | |
| 83982 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction