missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI27 Antibody (CL14627) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26876C
This item is not returnable.
View return policy
Description
IFI27 Monoclonal antibody specifically detects IFI27 in Human samples. It is validated for Immunohistochemistry
Specifications
| IFI27 | |
| Monoclonal | |
| DyLight 650 | |
| FAM14D, Interferon alpha-induced 11.5 kDa protein, interferon alpha-inducible protein 27, mitochondrial, interferon, alpha-inducible protein 27,2310061N23Rik, Interferon-stimulated gene 12a protein, ISG12, ISG12(a), ISG12A, P27 | |
| This antibody was developed using a synthetic peptide derived from P40305, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 3429 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry | |
| CL14627 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction