Learn More
IFI27 Antibody (CL14627) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26876AF647
Product Code. 30290242
  • £599.00 / 0.10mL
Description
IFI27 Monoclonal antibody specifically detects IFI27 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFI27
Monoclonal
Alexa Fluor 647
FAM14D, Interferon alpha-induced 11.5 kDa protein, interferon alpha-inducible protein 27, mitochondrial, interferon, alpha-inducible protein 27,2310061N23Rik, Interferon-stimulated gene 12a protein, ISG12, ISG12(a), ISG12A, P27
This antibody was developed using a synthetic peptide derived from P40305, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Cancer
3429
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry
CL14627
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified