missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ifi202 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523493
This item is not returnable.
View return policy
Description
Ifi202 Polyclonal Antibody for Western Blot
Interferon-activable protein 202 belongs to the HIN-200 family and contains two HIN-200 domains. It is reported to inhibit transcriptional activity of several transcription factors, including NF-kappaB p50 and p65, FOS, JUN, E2F1, E2F4, MYOD1 and myogenin. It has anti-apoptotic effects due to inhibition of the transcriptional activity of p53. It is an interferon-inducible protein which is responsible for Lupus susceptibility and is induced by Interleukin-6.
Specifications
| Ifi202 | |
| Polyclonal | |
| Unconjugated | |
| Ifi202b | |
| Ifbip-1; Ifi202; Ifi-202; Ifi202a; Ifi202b; interferon activated gene 202A; interferon activated gene 202B; interferon-activable protein 202; Interferon-activatable protein 202a (Ifi-202a) (Interferon-inducible protein p202a); interferon-inducible protein p202; Lupus susceptibility protein p202; p202 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100044068, 26388 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9R002 | |
| Ifi202b, LOC100044068 | |
| A synthetic peptide corresponding to amino acids 273-290 of the mouse Ifi202b protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction