missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFI16 Recombinant Mouse Monoclonal Antibody (K56050_2F2)

Product Code. 30830391
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830391 50 μL
30827096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830391 Supplier Invitrogen™ Supplier No. MA565928
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: EVDATSPAPS TSSTVKTEGA EATPGAQKRK KSTKEKAGPK GSKVSEEQTQ PPSPAGAGMS TAMGRSPSPK TSLSAPPNSS STENPKTVAK CQVTPRRNVL QKRPVIVKVL STTKPFEYET PEMEKKIMFH ATVATQTQFF HVKVLNTSLK EKFNGKKIII ISDYLEYDSL LEVNEESTVS EAGPNQTFEV PNKIINRAKE TLKIDILHKQ ASGNIVYGVF MLHKKTVNQK TTIYEIQDDR GKMDVVGTGQ CHNIPCEEGD KLQLFCFRLR KKNQMSKLIS EMHSFIQIKK KTNPRNNDPK SMKLPQEQRQ LPYPSEASTT FPESHLRTPQ MPPTTPSSSF Highest antigen sequence identity to the following orthologs: Mouse - 49%, Pig - 54%, Cynomolgus monkey - 89%.

IFI16 (gamma-inteferon-inducible protein 16) binds double-stranded DNA. It binds preferentially to supercoiled DNA and cruciform DNA structures. It seems to be involved in transcriptional regulation and as a transcriptional repressor. IFI16 may have a role in the regulation of hematopoietic differentiation through activation of unknown target genes. It controls cellular proliferation by modulating the functions of cell cycle regulatory factors including p53/TP53 and the retinoblastoma protein. IFI16 is also involved in innate immune response by recognizing viral dsDNA in the cytosol and nucleus. After binding to viral DNA in the cytoplasm, IFI16 recruits TMEM173/STING and mediates the induction of IFN-beta. It is necessary to activate the IFR3 signaling cascade suring human herpes simplex virus 1 infection and promotes the assembly of heterochromatin on herpesviral DNA and inhibiton of viral immediate-early gene expression and replication. IFI16 is also involved in the MTA1-mediated epigenetic regulation of ESR1 expression in breast cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI16
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K56050_2F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.09% sodium azide
Gene IFI16
Gene Accession No. Q16666
Gene Alias gamma-interferon-inducible protein 16; IFI 16C; IFI16; Ifi-16; IFNGIP1; interferon gamma inducible protein 16; interferon, gamma-inducible protein 16; interferon-gamma induced protein IFI 16; interferon-inducible myeloid differentiation transcriptional activator; PYHIN2
Gene Symbols IFI16
Host Species Mouse
Immunogen Recombinant human IFI16 (aa 101-440).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3428
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.