missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFI16 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £365.00
Specifications
| Antigen | IFI16 |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot, ELISA, Immunohistochemistry 1 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348864
|
Novus Biologicals
NBP3-18336-0.025MG |
0.025 mg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18346275
|
Novus Biologicals
NBP3-18336-0.1MG |
0.1 mg |
£365.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFI16 Polyclonal antibody specifically detects IFI16 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| IFI16 | |
| Western Blot, ELISA, Immunohistochemistry 1 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ifi-16, IFNGIP1gamma-interferon-inducible protein 16, interferon, gamma-inducible protein 16, interferon-gamma induced protein IFI 16, Interferon-inducible myeloid differentiation transcriptional activator, MGC9466, PYHIN2 | |
| Antibody was raised against a peptide corresponding to 16 amino acids near the carboxyl terminus of human IFI16. The immunogen is located within the last 50 amino acids of IFI16. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research | |
| PBS | |
| 3428 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title