missing translation for 'onlineSavingsMsg'
Learn More

IFI16 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18373797 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373797 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373797 Supplier Novus Biologicals Supplier No. NBP318336JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IFI16 Polyclonal antibody specifically detects IFI16 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFI16
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ifi-16, IFNGIP1gamma-interferon-inducible protein 16, interferon, gamma-inducible protein 16, interferon-gamma induced protein IFI 16, Interferon-inducible myeloid differentiation transcriptional activator, MGC9466, PYHIN2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the carboxyl terminus of human IFI16. The immunogen is located within the last 50 amino acids of IFI16.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 3428
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.