missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IFI16 Polyclonal antibody specifically detects IFI16 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | IFI16 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | PBS |
| Gene Alias | ifi-16, IFNGIP1gamma-interferon-inducible protein 16, interferon, gamma-inducible protein 16, interferon-gamma induced protein IFI 16, Interferon-inducible myeloid differentiation transcriptional activator, MGC9466, PYHIN2 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a peptide corresponding to 16 amino acids near the carboxyl terminus of human IFI16. The immunogen is located within the last 50 amino acids of IFI16. |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?