missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFFO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121114
This item is not returnable.
View return policy
Description
Positive test controls include: SKOV3, U-251MG.
IFFO2 is a protein coding gene.
Specifications
| IFFO2 | |
| Polyclonal | |
| Unconjugated | |
| IFFO2 | |
| IFFO2; Intermediate filament family orphan 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 126917, 212632, 641315 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P0C6R4, Q5TF58, Q8R2V2 | |
| IFFO2 | |
| Recombinant protein of human IFFO2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction