missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFFO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564413
This item is not returnable.
View return policy
Description
Immunogen sequence: LLWEDFSGYA MAAAEAQGEQ QEDSLEKVIK DTESLFKTR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| IFFO1 | |
| Polyclonal | |
| Unconjugated | |
| IFFO1 | |
| 4733401N06Rik; A930037G23Rik; HOM-TES-103; Iffo; IFFO1; Intermediate filament family orphan 1; intermediate filament-like MGC:2625; RGD1308257; Tumor antigen HOM-TES-103 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q0D2I5 | |
| IFFO1 | |
| Recombinant protein corresponding to Human IFFO1. Recombinant protein control fragment (Product #RP-104049). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction