missing translation for 'onlineSavingsMsg'
Learn More

Ifenprodil tartrate, MedChemExpress

Product Code. 16466026
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466026 100 mg 100mg
16411617 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16466026 Supplier Medchem Express Supplier No. HY12882A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ifenprodil tartrate is a typical noncompetitive NMDA receptor antagonist. Ifenprodil tartrate exerts high affinity at NR1A/NR2B receptors (IC50=0.34 μM) over 400-fold than at NR1A/NR2A receptors (IC50=146 μM). Ifenprodil tartrate inhibits GIRK (Kir3), reduces inward currents through the basal GIRK activity. Ifenprodil tartrate has the potential to be a cerebral vasodilator.

Ifenprodil tartrate is a typical noncompetitive NMDA receptor antagonist. Ifenprodil tartrate exerts high affinity at NR1A/NR2B receptors (IC50=0.34 μM) over 400-fold than at NR1A/NR2A receptors (IC50=146 μM). Ifenprodil tartrate inhibits GIRK (Kir3), reduces inward currents through the basal GIRK activity. Ifenprodil tartrate has the potential to be a cerebral vasodilator.

Specifications

Chemical Name or Material Ifenprodil tartrate
CAS 23210-58-4
Color White
Purity Grade Notes Research
Molecular Formula C21H27NO2.1/2C4H6O6
Quantity 100 mg
Solubility Information DMSO : ≥ 500 mg/mL (1248.47 mM) ∣H2O : 5 mg/mL (12.48 mM; Need ultrasonic)
SMILES OC(C1=CC=C(O)C=C1)C(C)N(CC2)CCC2CC3=CC=CC=C3.O=C(O)[C@H](O)[C@@H](O)C(O)=O.[1/2]
Molecular Weight (g/mol) 400.49
Formula Weight 400.49
Percent Purity 99.58%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.