missing translation for 'onlineSavingsMsg'
Learn More

Ifenprodil Hemitartrate, TRC

Product Code. 30396591
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
Unit Size:
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396591 10 mg 10mg
30403094 250 mg -
30405533 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396591 Supplier Toronto Research Chemicals Supplier No. TRCI25800010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23210-58-4
Molecular Formula 2 C21 H27 N O2 . C4 H6 O6
Molecular Weight (g/mol) 800.98
Synonym 1-Piperidineethanol, α-(4-hydroxyphenyl)-β-methyl-4-(phenylmethyl)-, (2R,3R)-2,3-dihydroxybutanedioate (2:1), 1-Piperidineethanol, 4-benzyl-α-(p-hydroxyphenyl)-β-methyl-, tartrate (salt) (8CI), 1-Piperidineethanol, α-(4-hydroxyphenyl)-β-methyl-4-(phenylmethyl)-, (2R,3R)-2,3-dihydroxybutanedioate (2:1) (salt) (9CI), 1-Piperidineethanol, α-(4-hydroxyphenyl)-β-methyl-4-(phenylmethyl)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (2:1) (salt), Ifenprodil L-(+)-tartrate, Ifenprodil tartrate
IUPAC Name 4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol;(2R,3R)-2,3-dihydroxybutanedioic acid
SMILES CC(C(O)c1ccc(O)cc1)N2CCC(Cc3ccccc3)CC2.CC(C(O)c4ccc(O)cc4)N5CCC(Cc6ccccc6)CC5.O[C@H]([C@@H](O)C(=O)O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/2C21H27NO2.C4H6O6/c2*1-16(21(24)19-7-9-20(23)10-8-19)22-13-11-18(12-14-22)15-17-5-3-2-4-6-17;5-1(3(7)8)2(6)4(9)10/h2*2-10,16,18,21,23-24H,11-15H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1
Formula Weight 800.4248
Chemical Name or Material Ifenprodil Hemitartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.