missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IER3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£150.00
Specifications
| Antigen | IER3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IER3 Polyclonal specifically detects IER3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| IER3 | |
| Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.02% Sodium Azide | |
| DIF-2, DIF2GLY96, Differentiation-dependent gene 2 protein, expressed in pancreatic carcinoma, gly96, mouse, homolog of, IEX-1, IEX1death protein, IEX-1L, Immediate early protein GLY96, immediate early response 3, Immediate early response 3 protein, immediately early gene X-1, PRG1PACAP-responsive gene 1 protein, Protein DIF-2, Protein PRG1, radiation-inducible immediate-early gene IEX-1 | |
| IER3 | |
| IgG | |
| Affinity Purified | |
| 17 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P46695 | |
| 8870 | |
| Antibody was raised against a 24 amino acid synthetic peptide near the center of human IEX1. The immunogen is located within amino acids 40 - 90 of IEX-1. Amino Acid Squence: APAGRPSASRGHRKRSRRVLYPR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel