missing translation for 'onlineSavingsMsg'
Learn More

IER3 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18407934 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407934 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407934 Supplier Novus Biologicals Supplier No. NBP176802AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IER3 Polyclonal antibody specifically detects IER3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IER3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias DIF-2, DIF2GLY96, Differentiation-dependent gene 2 protein, expressed in pancreatic carcinoma, gly96, mouse, homolog of, IEX-1, IEX1death protein, IEX-1L, Immediate early protein GLY96, immediate early response 3, Immediate early response 3 protein, immediately early gene X-1, PRG1PACAP-responsive gene 1 protein, Protein DIF-2, Protein PRG1, radiation-inducible immediate-early gene IEX-1
Host Species Rabbit
Immunogen Antibody was raised against a 24 amino acid synthetic peptide near the center of human IEX1. The immunogen is located within amino acids 40 - 90 of IEX-1. Amino Acid Squence: APAGRPSASRGHRKRSRRVLYPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8870
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.