missing translation for 'onlineSavingsMsg'
Learn More

IEICO-4Cl, Thermo Scientific Chemicals

Artikelnummer. 17196574
Ansicht ändern
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Packungsgröße:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
17196574 250 mg 250mg
17186574 100 mg 100mg
17106584 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17196574 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. H66752.MD
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2240998-88-1
Molecular Formula C{1}{1}{4}H{1}{1}{4}Cl{4}N{4}O{4}S{4}
Synonym 2,2 '-((2 Z,2 'Z)-(((4,4,9-tris(4-hexylphenyl)-9-(4-pentylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene5,2-diyl))bis(methanylylidene))bis(5,6-dichloro-3-oxo-2,3-dihydro-1 H-indene-2,1-diylidene))dimalononitrile

Specifications

Quantity 250 mg
Formula Weight 1874.22
Physical Form Solid
Chemical Name or Material IEICO-4 Cl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.