missing translation for 'onlineSavingsMsg'
Learn More

Iduronate 2-Sulfatase/IDS Antibody (331320) [DyLight 488], Novus Biologicals™

Product Code. 30733245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733245 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733245 Supplier Novus Biologicals Supplier No. FAB2449K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Iduronate 2-Sulfatase/IDS Monoclonal antibody specifically detects Iduronate 2-Sulfatase/IDS in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Iduronate 2-Sulfatase/IDS
Applications Western Blot
Classification Monoclonal
Clone 331320
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Iduronate 2-Sulfatase/IDS, Ser26-Pro550, Accession # P22304
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 3423
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.