missing translation for 'onlineSavingsMsg'
Learn More

IDRA-21, TRC

Product Code. 30398065
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398065 250 mg
30396061 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398065 Supplier Toronto Research Chemicals Supplier No. TRCI232500250MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22503-72-6
Molecular Formula C8H9ClN2O2S
Molecular Weight (g/mol) 232.69
Synonym 7-Chloro-3,4-dihydro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-Dioxide
IUPAC Name 7-chloro-3-methyl-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine 1,1-dioxide
SMILES CC1Nc2ccc(Cl)cc2S(=O)(=O)N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H9ClN2O2S/c1-5-10-7-3-2-6(9)4-8(7)14(12,13)11-5/h2-5,10-11H,1H3
Formula Weight 232.0073
Chemical Name or Material IDRA-21
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.