Learn More
Tocris Bioscience™ IDRA 21
Inhibits AMPA receptor desensitization
Specifications
Purity >98%
View More Specs
Description
Description
Inhibits AMPA receptor desensitization and enhances cognition by a related mechanism. More able to cross the blood-brain barrier than cyclothiazide (Cat. No. 0713).
Specifications
Specifications
>98%